Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBP4/Retinol-Binding Protein 4 Antibody (001), Alexa Fluor™ 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290460AF594
Description
RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
RBP4/Retinol-Binding Protein 4 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human RBP4 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
1 | |
Western Blot, ELISA | |
interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse RBP4/Retinol-Binding Protein 4 (NP_035385.1; Met 1-Leu 201). | |
0.1 mL | |
Cancer, Diabetes Research, Lipid and Metabolism | |
5950 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction