Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBP4/Retinol-Binding Protein 4 Antibody (9G7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | RBP4/Retinol-Binding Protein 4 |
---|---|
Clone | 9G7 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
RBP4/Retinol-Binding Protein 4 Monoclonal antibody specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
RBP4/Retinol-Binding Protein 4 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma | |
A synthesized peptide derived from Human RBP4/Retinol-Binding Protein 4 [UniProt P02753] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
9G7 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Diabetes Research, Lipid and Metabolism | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5950 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title