Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBP4/Retinol-Binding Protein 4 Rabbit anti-Rat, Clone: 3, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | RBP4/Retinol-Binding Protein 4 |
---|---|
Clone | 3 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Rat samples. It is validated for ELISA.Specifications
RBP4/Retinol-Binding Protein 4 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Rat | |
interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma | |
Recombinant Rat RBP4/Retinol-Binding Protein 4 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
3 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Diabetes Research, Lipid and Metabolism | |
0.2 um filtered solution in PBS | |
5950 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title