Learn More
RBP5 Antibody, HRP, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP297021H
Catalog No. NB080690
Only null left
Add to Cart
Description
Description
RBP5 Polyclonal specifically detects RBP5 in Human samples. It is validated for ELISA.
Specifications
Specifications
RBP5
Polyclonal
PBS
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA
HRP
Cellular retinol-binding protein III, CRBP3, CRBPIII, CRBP-III, HRBPiso, putative cellular retinol-binding protein CRBP III, retinol binding protein 5, cellular, retinol-binding protein 5, retinol-binding protein 5, cellular
Produced in rabbits immunized with purified, recombinant RBP5 (P82980; Met1-Arg135).
0.1 mL
Signal Transduction
83758
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.