Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104544 Supplier Invitrogen™ Supplier No. PA5104544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RBP5.

The protein encoded by this gene is a cellular retinol-binding protein expressed highly in kidney and liver. Down-regulation of the encoded protein in hepatocellular carcinoma was associated with large tumor size and poor patient survival rates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RBP5
Gene Accession No. P82980
Gene Alias cellular retinol binding protein 5; cellular retinol-binding protein III; CRBP3; CRBPIII; CRBP-III; HRBPiso; putative cellular retinol-binding protein CRBP III; RBP5; retinol binding protein 5; retinol binding protein 5, cellular; retinol-binding protein 5; retinol-binding protein 5, cellular
Gene Symbols RBP5
Host Species Rabbit
Immunogen A synthesized peptide derived from human RBP5(Accession P82980), corresponding to amino acid residues G80-V130.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83758
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.