Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106491
Description
Antibody detects endogenous levels of total RBP5.
The protein encoded by this gene is a cellular retinol-binding protein expressed highly in kidney and liver. Down-regulation of the encoded protein in hepatocellular carcinoma was associated with large tumor size and poor patient survival rates.
Specifications
| RBP5 | |
| Polyclonal | |
| Unconjugated | |
| RBP5 | |
| cellular retinol binding protein 5; cellular retinol-binding protein III; CRBP3; CRBPIII; CRBP-III; HRBPiso; putative cellular retinol-binding protein CRBP III; RBP5; retinol binding protein 5; retinol binding protein 5, cellular; retinol-binding protein 5; retinol-binding protein 5, cellular | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83758 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P82980 | |
| RBP5 | |
| A synthesized peptide derived from human RBP5(Accession P82980), corresponding to amino acid residues G80-V130. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction