Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP7 Monoclonal Antibody (4F4)
GREENER_CHOICE

Catalog No. PIMA524514
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524514 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524514 Supplier Invitrogen™ Supplier No. MA524514
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: KLLKPQKVIE QNGDSFTIHT NSSLRNYFVK FKVGEEFDED NRGLDNRKCK SLVIWDNDRL TCIQKGEKKN RGWTHWIEGD KLHLEMFCEG QVCKQTFQRA.

Due to its chemical instability and low solubility in aqueous solution, vitamin A requires cellular retinol-binding proteins (CRBPs), such as RBP7, for stability, internalization, intercellular transfer, homeostasis, and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4F4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Rbp7
Gene Accession No. Q96R05
Gene Alias 1110002J23Rik; cellular retinoic acid-binding protein 4; Cellular retinoic acid-binding protein IV; cellular retinol binding protein 7; cellular retinol binding protein type III; CRABP4; CRABP-IV; CRBP4; CRBP-III; CRBPIV; cytosolic binding protein; putative cellular retinol-binding protein CRBP IV; RBP7; retinoid binding protein 7; retinoid-binding protein 7; retinol binding protein 7; retinol binding protein 7, cellular; RGD1562168
Gene Symbols Rbp7
Host Species Mouse
Immunogen RBP7 (NP_443192.1, 35 a.a. ~ 134 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116362
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.