Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP7 Polyclonal Antibody

Catalog No. PIPA5121649
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121649 20 μL
PIPA5119393 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121649 Supplier Invitrogen™ Supplier No. PA5121649
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RBP7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Due to its chemical instability and low solubility in aqueous solution, vitamin A requires cellular retinol-binding proteins (CRBPs), such as RBP7, for stability, internalization, intercellular transfer, homeostasis, and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Rbp7
Gene Accession No. Q96R05
Gene Alias 1110002J23Rik; cellular retinoic acid-binding protein 4; Cellular retinoic acid-binding protein IV; cellular retinol binding protein 7; cellular retinol binding protein type III; CRABP4; CRABP-IV; CRBP4; CRBP-III; CRBPIV; cytosolic binding protein; putative cellular retinol-binding protein CRBP IV; RBP7; retinoid binding protein 7; retinoid-binding protein 7; retinol binding protein 7; retinol binding protein 7, cellular; RGD1562168
Gene Symbols Rbp7
Host Species Rabbit
Immunogen Recombinant protein Retinol Binding Protein 7, Cellular.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100270723, 116362
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.