Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590839 Supplier Invitrogen™ Supplier No. PA590839
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPADLSGTWT LLSSDNFEGY MLALGIDFAT RKIAKLLKPQ KVIEQNGDSF TIHTNSSLRN YFVKFKVGEE FDEDNRGLDN RKCKSLVIWD NDRLTCIQKG EKKNRGWTHW IEGDKLHLEM FCEGQVCKQT FQRA; Positive Samples: Mouse heart, Rat heart; Cellular Location: Cytoplasm.

Due to its chemical instability and low solubility in aqueous solution, vitamin A requires cellular retinol-binding proteins (CRBPs), such as RBP7, for stability, internalization, intercellular transfer, homeostasis, and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Rbp7
Gene Accession No. Q9EPC5
Gene Alias 1110002J23Rik; cellular retinoic acid-binding protein 4; Cellular retinoic acid-binding protein IV; cellular retinol binding protein 7; cellular retinol binding protein type III; CRABP4; CRABP-IV; CRBP4; CRBP-III; CRBPIV; cytosolic binding protein; putative cellular retinol-binding protein CRBP IV; RBP7; retinoid binding protein 7; retinoid-binding protein 7; retinol binding protein 7; retinol binding protein 7, cellular; RGD1562168
Gene Symbols Rbp7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-134 of human RBP7 (NP_4431921).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362662, 63954
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.