Learn More
RBPJ Monoclonal Antibody (1F1), Active Motif™

Description
Specifications
Specifications
| Antigen | RBPJ |
| Applications | ChIP Assay, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1F1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | RBPJ |
| Gene Accession No. | P31266, Q06330 |
| Gene Alias | AI843960; AOS3; BOS_6494; CBF1; CBF-1; csl; H-2K binding factor-2; IGKJRB; IGKJRB1; Igkrsbp; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; KBF2; LOC100346517; Rbpj; RBP-J; RBP-J kappa; RBPJK; RBP-JK; RBPSUH; recombination signal binding protein for immunoglobulin kappa J region; Recombining binding protein suppressor of hairless; recombining binding protein suppressor of hairless-like; Renal carcinoma antigen NY-REN-30; SUH; suppressor of hairless homolog |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.