Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBPJ Monoclonal Antibody (7A11), Active Motif™
SDP

Catalog No. 501993113
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3113 10 μg
50-199-3112 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3113 Supplier Active Motif Supplier No. 61074
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.

Specifications

Antigen RBPJ
Applications Western Blot
Classification Monoclonal
Clone 7A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene RBPJ
Gene Accession No. Q06330
Gene Alias AI843960; AOS3; BOS_6494; CBF1; CBF-1; csl; H-2K binding factor-2; IGKJRB; IGKJRB1; Igkrsbp; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; KBF2; LOC100346517; Rbpj; RBP-J; RBP-J kappa; RBPJK; RBP-JK; RBPSUH; recombination signal binding protein for immunoglobulin kappa J region; Recombining binding protein suppressor of hairless; recombining binding protein suppressor of hairless-like; Renal carcinoma antigen NY-REN-30; SUH; suppressor of hairless homolog
Gene Symbols RBPJ
Host Species Rat
Immunogen This antibody was raised against full-length recombinant human RBPJ protein.
Purification Method Protein G
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3516
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.