Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBPJ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569762 Supplier Invitrogen™ Supplier No. PA569762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 92%; Zebrafish: 92%.

CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBPJ
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBPJ
Gene Accession No. P31266
Gene Alias AI843960; AOS3; CBF1; CBF-1; csl; H-2K binding factor-2; Igkjrb; Igkjrb1; Igkrsbp; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; KBF2; Rbpj; RBP-J; RBP-J kappa; RBPJK; RBP-JK; rbp-jkappa; RBPSUH; recombination signal binding protein for immunoglobulin kappa J region; recombining binding protein suppressor of hairless; recombining binding protein suppressor of hairless-like; recombining binding protein suppressor of hairless-like protein; renal carcinoma antigen NY-REN-30; SUH; suppressor of hairless homolog; Unknown (protein for MGC:128782)
Gene Symbols RBPJ
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse RBPJ.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19664
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.