Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBPJ Polyclonal Antibody, Invitrogen™

Catalog No. PIPA540354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540354 Supplier Thermo Scientific Supplier No. PA540354

Rabbit Polyclonal Antibody

Peptide sequence: RPHCSAAGAI LRANSSQVPP NESNTNSEGS YTNASTNSTS VTSSTATVVS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%

RBPJ Polyclonal antibody specifically detects RBPJ in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBPJ
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RBPJ
Gene Accession No. P31266, Q06330
Gene Alias AI843960; AOS3; CBF1; CBF-1; csl; H-2K binding factor-2; Igkjrb; Igkjrb1; Igkrsbp; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; KBF2; Rbpj; RBP-J; RBP-J kappa; RBPJK; RBP-JK; rbp-jkappa; RBPSUH; recombination signal binding protein for immunoglobulin kappa J region; recombining binding protein suppressor of hairless; recombining binding protein suppressor of hairless-like; recombining binding protein suppressor of hairless-like protein; renal carcinoma antigen NY-REN-30; SUH; suppressor of hairless homolog; Unknown (protein for MGC:128782)
Gene Symbols RBPJ
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RBPSUH
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19664, 3516
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.