Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBPMS Antibody (HL1105), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP313722
This item is not returnable.
View return policy
Description
RBPMS Monoclonal antibody specifically detects RBPMS in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
RBPMS | |
Monoclonal | |
Unconjugated | |
FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing | |
Recombinant protein encompassing a sequence within the center region of human RBPMS. The exact sequence is proprietary. | |
100 μg | |
DNA Repair, Mismatch Repair | |
11030 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
HL1105 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction