Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBPMS Antibody (OTI2G10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273835F
Description
RBPMS Monoclonal specifically detects RBPMS in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
RBPMS | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
RBPMS | |
Full length recombinant protein of human RBPMS (NP_001008712) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry, Immunohistochemistry | |
OTI2G10 | |
Flow Cytometry, Immunohistochemistry | |
FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing | |
Mouse | |
Affinity Purified | |
DNA Repair, Mismatch Repair | |
11030.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction