Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBPMS Antibody (OTI3B7), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273836APC
Description
RBPMS Monoclonal specifically detects RBPMS in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.Specifications
| RBPMS | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| RBPMS | |
| Full length recombinant protein of human RBPMS (NP_001008712) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI3B7 | |
| Flow Cytometry | |
| FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing | |
| Mouse | |
| Affinity Purified | |
| DNA Repair, Mismatch Repair | |
| 11030.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction