Learn More
Invitrogen™ RBPMS Monoclonal Antibody (OTI2C3)

Description
RBPMS Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RBPMS |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2C3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Rbpms |
| Gene Accession No. | Q93062 |
| Gene Alias | 2010300K22Rik; 2700019M19Rik; AU017537; Heart and RRM expressed sequence; HEart, RRM Expressed Sequence; HERMES; Rbpms; RBP-MS; RGD1561067; RNA binding protein gene with multiple splicing; RNA binding protein with multiple splicing; RNA binding protein, mRNA processing factor; RNA-binding protein gene with multiple splicing; RNA-binding protein with multiple splicing; RNA-binding protein with multiple splicing (RBP-MS) |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.