Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBPMS Monoclonal Antibody (OTI3B7)
GREENER_CHOICE

Catalog No. PIMA526396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526396 Supplier Invitrogen™ Supplier No. MA526396
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBPMS Monoclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes a member of the RRM family of RNA-binding proteins. The RRM domain is between 80-100 amino acids in length and family members contain one to four copies of the domain. The RRM domain consists of two short stretches of conserved sequence called RNP1 and RNP2, as well as a few highly conserved hydrophobic residues. The protein encoded by this gene has a single, putative RRM domain in its N-terminus. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBPMS
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rbpms
Gene Accession No. Q93062
Gene Alias 2010300K22Rik; 2700019M19Rik; AU017537; Heart and RRM expressed sequence; HEart, RRM Expressed Sequence; HERMES; Rbpms; RBP-MS; RGD1561067; RNA binding protein gene with multiple splicing; RNA binding protein with multiple splicing; RNA binding protein, mRNA processing factor; RNA-binding protein gene with multiple splicing; RNA-binding protein with multiple splicing; RNA-binding protein with multiple splicing (RBP-MS)
Gene Symbols Rbpms
Host Species Mouse
Immunogen Full length human recombinant protein of RBPMS produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11030, 498642
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.