Learn More
RBPMS Mouse anti-Canine, Human, Clone: OTI4H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800216

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the RRM family of RNA-binding proteins. The RRM domain is between 80-100 amino acids in length and family members contain one to four copies of the domain. The RRM domain consists of two short stretches of conserved sequence called RNP1 and RNP2, as well as a few highly conserved hydrophobic residues. The protein encoded by this gene has a single, putative RRM domain in its N-terminus. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
RBPMS | |
Monoclonal | |
Unconjugated | |
RBPMS | |
HERMES | |
Mouse | |
Affinity Chromatography | |
RUO | |
11030, 475599 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI4H6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q93062 | |
RBPMS | |
Full length human recombit protein of human RBPMS produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.