Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBPMS Recombinant Rabbit Monoclonal Antibody (HL1105)
GREENER_CHOICE

Catalog No. PIMA542376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542376 Supplier Invitrogen™ Supplier No. MA542376
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

This gene encodes a member of the RRM family of RNA-binding proteins. The RRM domain is between 80-100 amino acids in length and family members contain one to four copies of the domain. The RRM domain consists of two short stretches of conserved sequence called RNP1 and RNP2, as well as a few highly conserved hydrophobic residues. The protein encoded by this gene has a single, putative RRM domain in its N-terminus. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBPMS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL1105
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Rbpms
Gene Accession No. Q93062, Q9WVB0
Gene Alias 2010300K22Rik; 2700019M19Rik; AU017537; Heart and RRM expressed sequence; HEart, RRM Expressed Sequence; HERMES; Rbpms; RBP-MS; RGD1561067; RNA binding protein gene with multiple splicing; RNA binding protein with multiple splicing; RNA binding protein, mRNA processing factor; RNA-binding protein gene with multiple splicing; RNA-binding protein with multiple splicing; RNA-binding protein with multiple splicing (RBP-MS)
Gene Symbols Rbpms
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RBPMS. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11030, 19663, 498642
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.