Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBPSUHL Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984372
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4372 Supplier Bioss Supplier No. BS19783R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In mouse, recombining binding protein L (RBP-L) is a transcription factor that binds to DNA sequences almost identical to that bound by the Notch receptor signaling pathway transcription factor RBP-J. However, unlike RBP-J, RBP-L does not interact with Notch receptors. RBP-L has been shown to activate transcription in concert with Epstein-Barr virus nuclear antigen-2 (EBNA2). The protein encoded by this gene is similar in sequence to the mouse RPB-L protein and Drosophila suppressor of hairless protein.

Specifications

Antigen RBPSUHL
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RBPJL
Gene Accession No. Q9UBG7
Gene Alias RBP-J kappa-like; Rbpjl; Rbpl; RBP-L; Rbpsuhl; recombination signal binding protein for immunoglobulin kappa J region like; recombination signal binding protein for immunoglobulin kappa J region-like; Recombining binding protein suppressor of hairless-like protein; SUHL; Transcription factor RBP-L
Gene Symbols RBPJL
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RBPSUHL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11317
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.