Learn More
Thermo Scientific™ RBS™ 35 Concentrate, 1 kg PROMO
Description
Removes both organic and inorganic materials and metal ions.
Highlights:
- Ideal for the removal of difficult substances such as protein and blood serum residues, tars, distillation residues, silicone, and oils
- Suitable for radiochemical decontamination
- Moderately alkaline mixture of anionic and nonionic surfactants
- Safely and effectively cleans glass, plastic, quartz, porcelain, and ferrous metal
- Free-rinsing and biodegradable
Specifications
Specifications
| Form | Liquid |
| Product Line | RBS |
| Composition | 2% (v/v) Solution |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. |
| Quantity | 1 kg |
| Volume (Metric) | 1 kg |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.