Learn More
Thermo Scientific™ RBS™ 35 Concentrate, 20 kg PROMO
Catalog No. FER27852PM
Click to view available options
:
Thermo Scientific™ RBS™ 35 Concentrate
A no-residue laboratory cleaning solution for labware utensils, surface materials and glassware, including bottles, beakers, pipets, lenses, mirrors.
Removes both organic and inorganic materials and metal ions.
Highlights:
  • Ideal for the removal of difficult substances such as protein and blood serum residues, tars, distillation residues, silicone, and oils
  • Suitable for radiochemical decontamination
  • Moderately alkaline mixture of anionic and nonionic surfactants
  • Safely and effectively cleans glass, plastic, quartz, porcelain, and ferrous metal
  • Free-rinsing and biodegradable
Specifications
Quantity 20 kg
Physical Form Liquid
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.