Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RBS™ 35 Concentrate, 5 kg PROMO

Catalog No. PI27952PM
Click to view available options
Volume (Metric):
5 kg

Catalog No. PI27952PM

Supplier: Thermo Scientific™ 27952

A no-residue laboratory cleaning solution for labware utensils, surface materials and glassware, including bottles, beakers, pipets, lenses, mirrors.

Removes both organic and inorganic materials and metal ions.

Highlights:

  • Ideal for the removal of difficult substances such as protein and blood serum residues, tars, distillation residues, silicone, and oils
  • Suitable for radiochemical decontamination
  • Moderately alkaline mixture of anionic and nonionic surfactants
  • Safely and effectively cleans glass, plastic, quartz, porcelain, and ferrous metal
  • Free-rinsing and biodegradable

Specifications

Form Liquid
Product Line RBS
Composition 2% (v/v) Solution
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Quantity 5 kg
Volume (Metric) 5 kg

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.