Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ RBS™ Solid Detergent PROMO
Click to view available options
Volume (Metric):
5 kg
Description
Highlights:
- For automatic laboratory washing machines Low-foaming
- Rinses easily without leaving residues
- Fully biodegradable
Specifications
Specifications
Form | Powder |
Product Line | RBS™ |
Composition | Anionic and Nonionic Detergent, 2% (v/v) Concentrate Solution, 0.2% (w/v) Solid Solution |
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. |
Quantity | 5 kg |
Volume (Metric) | 5 kg |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction