Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RC3H1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585598
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585598 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585598 Supplier Invitrogen™ Supplier No. PA585598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (91%). Positive Control: 293T, A431, HeLa, HepG2, 3xFlag-human RC3H1 transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The ring finger protein RC3H1, also known as Roquin, is a highly conserved member of the RING type ubiquitin ligase protein family whose M199R mutation leads to the excessive production of follicular helper T cells and germinal centers in the sanroque strain of mice, a strain with excessive IL-21 production and high titers of autoantibodies. The complete loss of RC3H1 induces early death and immune deregulation but not autoimmunity in RC3H1-null mice, suggesting that the mutant RC3H1 is more disruptive to the immune system than its complete loss.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RC3H1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RC3H1
Gene Accession No. Q5TC82
Gene Alias 5730557L09Rik; Gm551; Kiaa2025; mKIAA2025; N28103; probable E3 ubiquitin-protein ligase Roquin; protein Sanroque; RC3H1; RGD1563512; RING CCCH (C3H) domains 1; RING finger and C3H zinc finger protein 1; ring finger and CCCH-type domains 1; RING finger and CCCH-type zinc finger domain-containing protein 1; ring finger and CCCH-type zinc finger domains 1; RING finger protein 198; RNF198; ROQUIN; Roquin-1
Gene Symbols RC3H1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human RC3H1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149041
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.