Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RC3H1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534519
Description
A suggested positive control is Hela cell lysate. PA5-34519 can be used with blocking peptide PEP-1562.
The ring finger protein RC3H1, also known as Roquin, is a highly conserved member of the RING type ubiquitin ligase protein family whose M199R mutation leads to the excessive production of follicular helper T cells and germinal centers in the sanroque strain of mice, a strain with excessive IL-21 production and high titers of autoantibodies. The complete loss of RC3H1 induces early death and immune deregulation but not autoimmunity in RC3H1-null mice, suggesting that the mutant RC3H1 is more disruptive to the immune system than its complete loss.
Specifications
| RC3H1 | |
| Polyclonal | |
| Unconjugated | |
| RC3H1 | |
| 5730557L09Rik; Gm551; Kiaa2025; mKIAA2025; N28103; probable E3 ubiquitin-protein ligase Roquin; protein Sanroque; RC3H1; RGD1563512; RING CCCH (C3H) domains 1; RING finger and C3H zinc finger protein 1; ring finger and CCCH-type domains 1; RING finger and CCCH-type zinc finger domain-containing protein 1; ring finger and CCCH-type zinc finger domains 1; RING finger protein 198; RNF198; ROQUIN; Roquin-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 149041, 381305, 680586 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q4VGL6, Q5TC82 | |
| RC3H1 | |
| An 18 amino acid peptide near the carboxy terminus of human RC3H1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction