Learn More
RC3H2/MNAB Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305150A

Description
The recommended shelf life for this product is 1 year from date of receipt.
RC3H2 gene ontology annotations related to this gene include B cell homeostasis; limb development; lung alveolus development; lymph node development; multicellular organism growth; negative regulation of T-helper 17 cell differentiation; positive regulation of NIK/NF-kappaB signaling; post-embryonic development; posttranscriptional regulation of gene expression; regulation of miRNA metabolic process; spleen development; T cell homeostasis; T cell proliferation; T cell receptor signaling pathway; T follicular helper cell differentiation.Specifications
RC3H2/MNAB | |
Polyclonal | |
Unconjugated | |
RC3H2 | |
FLJ20301, FLJ20713, membrane associated DNA binding protein, membrane-associated nucleic acid binding protein, membrane-associated nucleic acid-binding protein, MNAB, ring finger and CCCH-type zinc finger domain-containing protein 2, ring finger and CCCH-type zinc finger domains 2, RING finger protein 164, RNF164 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54542 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9HBD1 | |
RC3H2 | |
Between 1100 and 1150 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.