Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RC3H2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584534
Description
Immunogen sequence: QKEPPKQKKQ SLGEDHVILE EQKTILPVTS CFSQPLPVSI SNASCLPITT SVSAGNLILK THVMSEDKND FLKPVANGKM V.
RC3H2 gene ontology annotations related to this gene include B cell homeostasis; limb development; lung alveolus development; lymph node development; multicellular organism growth; negative regulation of T-helper 17 cell differentiation; positive regulation of NIK/NF-kappaB signaling; post-embryonic development; posttranscriptional regulation of gene expression; regulation of miRNA metabolic process; spleen development; T cell homeostasis; T cell proliferation; T cell receptor signaling pathway; T follicular helper cell differentiation.
Specifications
RC3H2 | |
Polyclonal | |
Unconjugated | |
RC3H2 | |
FLJ20301; FLJ20713; membrane associated DNA binding protein; membrane-associated nucleic acid binding protein; membrane-associated nucleic acid-binding protein; MNAB; RC3H2; ring finger and CCCH-type domains 2; ring finger and CCCH-type zinc finger domain-containing protein 2; ring finger and CCCH-type zinc finger domains 2; RING finger protein 164; RING-type E3 ubiquitin transferase Roquin-2; RNF164; Roquin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54542 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBD1 | |
RC3H2 | |
Recombinant protein corresponding to Human RC3H2. Recombinant protein control fragment (Product #RP-108466). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction