Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ VACUUBRAND™ RC6 Chemistry-HYBRID Vacuum Pump

For fine-vacuum applications

Specifications

Horsepower 0.5 hp
Model RC6
Noise Level 50 dbA
Temperature (Metric) Storage -10°C to +60°C
Weight (Metric) 24.2 kg
View More Specs

Products 1
Catalog Number Mfr. No. Plug Type Price Quantity & Availability  
Catalog Number Mfr. No. Plug Type Price Quantity & Availability  
13-689-411
View Documents
BrandTech™ VACUUBRAND™
20698563
US N/A N/A N/A
Description

Description

Combines rotary vane and diaphragm vacuum pump technology to support freeze drying and molecular distillations, with substantially reduced maintenance and less hazardous oil waste. Chemistry-design diaphragm pump is used to back the rotary vane pump. This configuration keeps pump oil under vacuum, reducing condensation of solvent vapors in pump oil and oil changes by as much as 90% over conventional rotary vane designs.

  • Free air capacity: 4.1cfm
  • Ultimate vacuum: 4 x 10-4mbar
  • Restriction-free, high-flow inlet ensures high flowrates under vacuum
  • Includes integrated oil mist filter

Order Info

Dimensions: 20L x 12W x9 in. H (51 x 30.5 x 23cm). Net weight: 53 lb. (24.2kg).

Specifications

Specifications

0.5 hp
50 dbA
24.2 kg
40°C
3.5/4.1 CFM
20 in.
IP40
RC6
-10°C to +60°C
104°F
12°C
50/60 Hz
12 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.