Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RC74 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565677
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565677 Supplier Invitrogen™ Supplier No. PA565677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAEWLCHNKQS KVYLPEPPFP HAELIQTNKL KHYPSIHGDF SNDFRQPCVV FTGHPSLRFG DVVHFMELWG KSSLNTVIFT EPDFSYLEAL AP Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1)and U2 (RNU2)(Baillat et al., 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RC74
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INTS9
Gene Accession No. Q9NV88
Gene Alias BC028953; CPSF2L; D14Ertd231e; INT9; integrator complex subunit 9; INTS9; Protein related to CPSF subunits of 74 kDa; RC74; RC-74
Gene Symbols INTS9
Host Species Rabbit
Immunogen Recombinant Human RC74. Recombinant protein control fragment (Product #RP-104929).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55756
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.