Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RC74 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566872
Description
Immunogen sequence: QPTSGKKRKRV SDDVPDCKVL KPLLSGSIPV EQFVQTLEKH GFSDIKVEDT AKGHIVLLQE AETLIQIEED STHIICDNDE MLRVRLRDLV LKFLQKF Highest antigen sequence identity to the following orthologs - mouse 94%, rat 95%.
INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1)and U2 (RNU2)(Baillat et al., 2005).
Specifications
RC74 | |
Polyclonal | |
Unconjugated | |
INTS9 | |
BC028953; CPSF2L; D14Ertd231e; INT9; integrator complex subunit 9; INTS9; Protein related to CPSF subunits of 74 kDa; RC74; RC-74 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55756 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NV88 | |
INTS9 | |
Recombinant Human RC74. Recombinant protein control fragment (Product #RP-105407). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction