Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RC74 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115433
Description
Antibody detects endogenous levels of total INTS9.
INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1)and U2 (RNU2)(Baillat et al., 2005).
Specifications
RC74 | |
Polyclonal | |
Unconjugated | |
INTS9 | |
BC028953; CPSF2L; D14Ertd231e; INT9; integrator complex subunit 9; INTS9; Protein related to CPSF subunits of 74 kDa; RC74; RC-74 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55756 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q9NV88 | |
INTS9 | |
A synthesized peptide derived from human INTS9(Accession Q9NV88), corresponding to amino acid residues P360-E410. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction