Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RCAN1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298713JF549
Description
RCAN1 Polyclonal antibody specifically detects RCAN1 in Human samples. It is validated for Western BlotSpecifications
RCAN1 | |
Polyclonal | |
Western Blot | |
ADAPT78, calcipressin-1, calcium and oxidant-inducible mRNA, Down syndrome candidate region 1, Down syndrome critical region gene 1, Down syndrome critical region protein 1, DSC1, modulatory calcineurin-interacting protein 1, Myocyte-enriched calcineurin-interacting protein 1, near DSCR proline-rich protein, regulator of calcineurin 1CSP1DSCR1MCIP1RCN1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RCAN1. | |
0.1 mL | |
Cancer, Neuroscience, Signal Transduction | |
1827 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction