Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RCAN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323371
Description
RCAN1 Polyclonal antibody specifically detects RCAN1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
RCAN1 | |
Polyclonal | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
ADAPT78, calcipressin-1, calcium and oxidant-inducible mRNA, Down syndrome candidate region 1, Down syndrome critical region gene 1, Down syndrome critical region protein 1, DSC1, modulatory calcineurin-interacting protein 1, Myocyte-enriched calcineurin-interacting protein 1, near DSCR proline-rich protein, regulator of calcineurin 1CSP1DSCR1MCIP1RCN1 | |
RCAN1 (C-terminus) synthetic peptide (coupled to carrier protein) corresponds to amino acids in the C-terminus of human RCAN1. This sequence is highly conserved in rat and mouse RCAN1, and this sequence is found in all four isoforms of RCAN1, but is not found in RCAN2 or RCAN3. [UniProt# P53805] | |
0.1 mL | |
Cancer, Neuroscience, Signal Transduction | |
1827 | |
Store at -20°C. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction