Learn More
Invitrogen™ RCAN1 Polyclonal Antibody

Description
RCAN1 Polyclonal Antibody detects endogenous levels of RCAN1 protein.
Specifications
Specifications
| Antigen | RCAN1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | Rcan1 |
| Gene Accession No. | P53805, Q6IN33, Q9JHG6 |
| Gene Alias | Adapt78; calcineurin inhibitor; calcipressin-1; calcium and oxidant-inducible mRNA; CALP1L; CSP1; Down syndrome candidate region 1; Down syndrome critical region gene 1; Down syndrome critical region homolog 1; Down syndrome critical region protein 1; Down syndrome critical region protein 1 homolog; DSC1; Dscr1; MCIP1; modulatory calcineurin-interacting protein 1; myocyte-enriched calcineurin-interacting protein 1; near DSCR proline-rich protein; Rcan1; RCN1; Regulator of calcineurin 1 |
| Gene Symbols | Rcan1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.