Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCAN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140242 Supplier Invitrogen™ Supplier No. PA5140242
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QEKEEEAEME RMRRPKPKII Q Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 76%.

The RCAN1 protein interacts with calcineurin A and inhibits calcineurin-dependent signaling pathways, possibly affecting central nervous system development. This gene is located in the minimal candidate region for the Down syndrome phenotype, and is overexpressed in the brain of Down syndrome fetuses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCAN1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rcan1
Gene Accession No. P53805
Gene Alias Adapt78; calcineurin inhibitor; calcipressin-1; calcium and oxidant-inducible mRNA; CALP1L; CSP1; Down syndrome candidate region 1; Down syndrome critical region gene 1; Down syndrome critical region homolog 1; Down syndrome critical region protein 1; Down syndrome critical region protein 1 homolog; DSC1; Dscr1; MCIP1; modulatory calcineurin-interacting protein 1; myocyte-enriched calcineurin-interacting protein 1; near DSCR proline-rich protein; Rcan1; RCN1; Regulator of calcineurin 1
Gene Symbols Rcan1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RCAN1. Recombinant protein control fragment (Product #RP-109156).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1827
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.