Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCAN2 Monoclonal Antibody (1A9)
GREENER_CHOICE

Catalog No. PIMA524549
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524549 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524549 Supplier Invitrogen™ Supplier No. MA524549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: KAEELHYPLG ERRSDYDREA LLGVQEDVDE YVKLGHEEQQ KRLQAIIKKI DLDSDGFLTE SELSSWIQMS FKHYAMQEAK QQFVEYDKNS DDTVTWDEYN IQMYDRVIDF DENTALDDAE EESFRKLHLK DKKRFEKANQ DSGPGLSLEE FIAFEHPEEV DYMTEFVIQE ALEEHDKNGD GFVSLEEFLG DYRWDPTANE DPEWILVEKD RFVNDYDKDN DGRLDPQELL PWVVPNNQGI AQEEALHLID EMDLNGDKKL SEEEILENPD LFLTSEATDY GRQLHDDYFY HDEL.

Regulator of calcineurin 2 (RCAN2), also known as ZAKI4 and DSCR1L1, is expressed as two isoforms differing at their N-terminus. The longer of the two (isoform 1) is expressed exclusively in the brain, while isoform 2 is ubiquitously expressed, with highest expression in brain, heart, and muscle. Both isoforms bind to the catalytic subunit of calcineurin, a Ca++-dependent protein phosphatase involved in several neuronal functions, though their C-terminal region and inhibit calcineurin's activity. Unlike isoform 1 of RCAN2, the expression of the second isoform is not induced by the thyroid hormone T3. RCAN2 is a member of a family of three endogenous calcineurin regulators that are located near the minimal supernumerary fragment of chromosome 21 in individuals with Down syndrome, suggesting that they play a role in this syndrome. Multiple isoforms of RCAN2 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCAN2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A9
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Rcan2
Gene Accession No. Q14206
Gene Alias calcineurin inhibitory protein ZAKI-4; Calcipressin-2; caskin-1-like; CSP2; Dcip2; Down syndrome candidate region 1-like 1; down syndrome candidate region 1-like protein 1; Down syndrome critical region gene 1-like 1; Down syndrome critical region gene 1-like 1 protein; Down syndrome critical region gene1-like 1; DSCR1L1; DSCR1L1 protein; LOW QUALITY PROTEIN: calcipressin-2; MCIP2; myocyte-enriched calcineurin interactin protein 2; myocyte-enriched calcineurin-interacting protein 2; Rcan2; RCN2; regulator of calcineurin 2; si:ch211-124g24.1; thyroid hormone-responsive (skin fibroblasts); thyroid hormone-responsive protein ZAKI-4; translation initiation factor IF-2; ZAKI4; ZAKI-4; zgc:85746
Gene Symbols Rcan2
Host Species Mouse
Immunogen RCN2 (AAH04892.1, 24 a.a. ~ 317 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10231
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.