Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RCAN2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572458
Description
Regulator of calcineurin 2 (RCAN2), also known as ZAKI4 and DSCR1L1, is expressed as two isoforms differing at their N-terminus. The longer of the two (isoform 1) is expressed exclusively in the brain, while isoform 2 is ubiquitously expressed, with highest expression in brain, heart, and muscle. Both isoforms bind to the catalytic subunit of calcineurin, a Ca++-dependent protein phosphatase involved in several neuronal functions, though their C-terminal region and inhibit calcineurin′s activity. Unlike isoform 1 of RCAN2, the expression of the second isoform is not induced by the thyroid hormone T3. RCAN2 is a member of a family of three endogenous calcineurin regulators that are located near the minimal supernumerary fragment of chromosome 21 in individuals with Down syndrome, suggesting that they play a role in this syndrome. Multiple isoforms of RCAN2 are known to exist.
Specifications
RCAN2 | |
Polyclonal | |
Unconjugated | |
RCAN2 | |
Csp2; Dscr1l1; MCIP2; ZAKI-4; Down syndrome critical region gene 1-like 1; calcineurin inhibitory protein ZAKI-4; calcipressin-2; down syndrome candidate region 1-like protein 1; myocyte-enriched calcineurin interactin protein 2; myocyte-enriched calcineurin-interacting protein 2 | |
Rabbit | |
Protein A | |
RUO | |
10231, 53901 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide | |
Q14206, Q9JHG2 | |
RCAN2 | |
KLH conjugated synthetic peptide between 6-37 amino acids from the N-terminal region of human DSCR1L1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction