Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCAN3 Monoclonal Antibody (OTI1B2), TrueMAB™, OriGene
SDP

Catalog No. 502539667
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9667 30 μL
50-167-5962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9667 Supplier OriGene Supplier No. TA505496S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RCAN3 Monoclonal antibody specifically detects RCAN3 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen RCAN3
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GADD45G
Gene Accession No. O95257
Gene Alias AU041093; Calcipressin 3; Calcipressin-3; Csp3; Down syndrome candidate region 1-like 2; down syndrome candidate region 1-like protein 2; Down syndrome critical region gene 1-like 2; DSCR1L2; dscr1l2 protein; hRCN3; MCIP3; myocyte-enriched calcineurin-interacting protein 3; RCAN family member 3; RCAN3; RCN3; Regulator of calcineurin 3; regulator of calcineurin 3 isoform 1b,2; regulator of calcineurin family member 3; similar to Calcipressin-3 (Down syndrome candidate region 1-like protein 2) (Myocyte-enriched calcineurin interacting protein 3) (MCIP3); wu:fc68f05; zgc:113155; zgc:91820
Host Species Mouse
Immunogen Full length human recombinant protein of human GADD45G produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10912
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.