Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCAN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541841 Supplier Invitrogen™ Supplier No. PA541841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PGEKYELHAG TESTPSVVVH VCESETEEEE ETKNPKQKIA QTRRPDPPTA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.

DSCR1L2 inhibits calcineurin-dependent transcriptional responses by binding to the catalytic domain of calcineurin A, and may play a role during central nervous system development. Highest expression occurs in heart, skeletal muscle kidney, liver and peripheral blood leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCAN3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RCAN3
Gene Accession No. Q9UKA8
Gene Alias AU041093; Calcipressin 3; Calcipressin-3; Csp3; Down syndrome candidate region 1-like 2; down syndrome candidate region 1-like protein 2; Down syndrome critical region gene 1-like 2; DSCR1L2; dscr1l2 protein; hRCN3; MCIP3; myocyte-enriched calcineurin-interacting protein 3; RCAN family member 3; RCAN3; RCN3; Regulator of calcineurin 3; regulator of calcineurin 3 isoform 1b,2; regulator of calcineurin family member 3; similar to Calcipressin-3 (Down syndrome candidate region 1-like protein 2) (Myocyte-enriched calcineurin interacting protein 3) (MCIP3); wu:fc68f05; zgc:113155; zgc:91820
Gene Symbols RCAN3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RCAN3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11123
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.