Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RCAN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556966
Description
Immunogen sequence: LRDTMKSWND SQSDLCSTDQ EEEEEMIFGE NEDDLDEMMD LSDLPTSLFA CSVHEAVFEA REQKERFEAL FTIYDD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
DSCR1L2 inhibits calcineurin-dependent transcriptional responses by binding to the catalytic domain of calcineurin A, and may play a role during central nervous system development. Highest expression occurs in heart, skeletal muscle kidney, liver and peripheral blood leukocytes.
Specifications
RCAN3 | |
Polyclonal | |
Unconjugated | |
RCAN3 | |
AU041093; Calcipressin 3; Calcipressin-3; Csp3; Down syndrome candidate region 1-like 2; down syndrome candidate region 1-like protein 2; Down syndrome critical region gene 1-like 2; DSCR1L2; dscr1l2 protein; hRCN3; MCIP3; myocyte-enriched calcineurin-interacting protein 3; RCAN family member 3; RCAN3; RCN3; Regulator of calcineurin 3; regulator of calcineurin 3 isoform 1b,2; regulator of calcineurin family member 3; similar to Calcipressin-3 (Down syndrome candidate region 1-like protein 2) (Myocyte-enriched calcineurin interacting protein 3) (MCIP3); wu:fc68f05; zgc:113155; zgc:91820 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKA8 | |
RCAN3 | |
Recombinant protein corresponding to Human RCAN3. Recombinant protein control fragment (Product #RP-94153). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction