Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCAS1 Monoclonal Antibody (01)
GREENER_CHOICE

Catalog No. PIMA530350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530350 Supplier Invitrogen™ Supplier No. MA530350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human RCAS1/EBAG9.

This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5'-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5' UTR, but encoding the same protein, have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCAS1
Applications ELISA
Classification Monoclonal
Clone 01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene EBAG9
Gene Accession No. O00559
Gene Alias AI835379; BAG9; cancer associated surface antigen; cancer associated surface antigen RCAS1; cancer-associated surface antigen RCAS1; EB9; EBAG9; estrogen receptor binding fragment associated gene 9; estrogen receptor binding site associated, antigen, 9; estrogen receptor-binding fragment-associated gene 9; estrogen receptor-binding fragment-associated gene 9 protein; HGNC:3123; PDAF; Rcas1; receptor binding cancer antigen expressed on SiSo cells; Receptor-binding cancer antigen expressed on SiSo cells
Gene Symbols EBAG9
Host Species Mouse
Immunogen Recombinant Human RCAS1/EBAG9 Protein (Arg28-Ser213).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9166
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.