Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCAS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102523 Supplier Invitrogen™ Supplier No. PA5102523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EBAG9.

This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5'-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5' UTR, but encoding the same protein, have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCAS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EBAG9
Gene Accession No. O00559, Q5PQP2, Q9D0V7
Gene Alias AI835379; BAG9; cancer associated surface antigen; cancer associated surface antigen RCAS1; cancer-associated surface antigen RCAS1; EB9; EBAG9; estrogen receptor binding fragment associated gene 9; estrogen receptor binding site associated, antigen, 9; estrogen receptor-binding fragment-associated gene 9; estrogen receptor-binding fragment-associated gene 9 protein; HGNC:3123; PDAF; Rcas1; receptor binding cancer antigen expressed on SiSo cells; Receptor-binding cancer antigen expressed on SiSo cells
Gene Symbols EBAG9
Host Species Rabbit
Immunogen A synthesized peptide derived from human EBAG9(Accession O00559), corresponding to amino acid residues Q175-S213.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299864, 55960, 9166
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.