Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RCBTB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530672 Supplier Invitrogen™ Supplier No. PA530672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, mouse brain. Predicted reactivity: Human (99%), Mouse (93%), Rat (93%), Rhesus Monkey (97%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein with an N-terminal RCC1 domain and a C-terminal BTB (broad complex, tramtrack and bric-a-brac) domain. In rat, over-expression of this gene in vascular smooth muscle cells induced cellular hypertrophy. In rat, the C-terminus of RCBTB1 interacts with the angiotensin II receptor-1A. In humans, this gene maps to a region of chromosome 13q that is frequently deleted in B-cell chronic lymphocytic leukemia and other lymphoid malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RCBTB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RCBTB1
Gene Accession No. Q6NXM2, Q8NDN9
Gene Alias 5430409I18Rik; AW111883; Chronic lymphocytic leukemia deletion region gene 7 protein; CLL deletion region gene 7 protein; CLLD7; CLLL7; E4.5; GDP/GTP exchange factor (GEF)-like protein; GLP; RCBTB1; RCC1 and BTB domain containing protein 1; RCC1 and BTB domain-containing protein 1; regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1; regulator of chromosome condensation and BTB domain-containing protein 1; RP11-185C18.1
Gene Symbols RCBTB1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RCBTB1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55213, 71330
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.