Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCBTB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599183 Supplier Thermo Scientific Supplier No. PA599183
Add to Cart
Add to Cart

RCBTB1 Polyclonal Antibody

RCBTB1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen RCBTB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RCBTB1
Gene Accession No. Q6NXM2, Q8NDN9
Gene Alias 5430409I18Rik; AW111883; Chronic lymphocytic leukemia deletion region gene 7 protein; CLL deletion region gene 7 protein; CLLD7; CLLL7; E4.5; GDP/GTP exchange factor (GEF)-like protein; GLP; RCBTB1; RCC1 and BTB domain containing protein 1; RCC1 and BTB domain-containing protein 1; regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1; regulator of chromosome condensation and BTB domain-containing protein 1; RP11-185C18.1
Gene Symbols RCBTB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RCBTB1(Accession Q8NDN9), corresponding to amino acid residues W257-V307.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55213, 71330
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.