Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RCBTB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112684
Description
RCBTB2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
RCBTB2 is a member of the RCC1-related GEF family. The N-terminal half of the amino acid sequence shows similarity to the regulator of chromosome condensation RCC1, which acts as a guanine nucleotide exchange factor (GEF) protein for the Ras-related GTPase Ran.
Specifications
| RCBTB2 | |
| Polyclonal | |
| Unconjugated | |
| Rcbtb2 | |
| 2610028E02Rik; 2810420M18Rik; AI448345; AW240694; CHC1L; CHC1-L; Chromosome condensation 1-like; RC/BTB2; Rcbtb2; RCC1 and BTB domain containing protein 2; RCC1 and BTB domain-containing protein 2; RCC1-like G exchanging factor; RCC1-like G exchanging factor RLG; regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2; regulator of chromosome condensation and BTB domain-containing protein 2; RLG | |
| Rabbit | |
| Protein G | |
| RUO | |
| 1102, 290363 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O95199, Q6P798 | |
| Rcbtb2 | |
| Recombinant Human RCC1 and BTB domain-containing protein 2 protein (1-200AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction