Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RCC1 Antibody (3D11), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197543AF594
Description
RCC1 Monoclonal specifically detects RCC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RCC1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
RCC1 | |
Recombinant human RanGEF. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
3D11 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Cell cycle regulatory protein, CHC1RCC1-I, chromosome condensation 1, Chromosome condensation protein 1, guanine nucleotide-releasing protein, regulator of chromosome condensation, regulator of chromosome condensation 1, SNHG3-RCC1, SNHG3-RCC1 readthrough transcript | |
Mouse | |
Protein A purified | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
1104.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction