Learn More
RCC1 Monoclonal Antibody (332CT2.1.1)

Description
RCC1 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | RCC1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 332CT2.1.1 |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RCC1 |
| Gene Accession No. | P18754, Q8VE37 |
| Gene Alias | 4931417M11Rik; AI326872; Cell cycle regulatory protein; chc1; chc1-a; chromosome condensation 1; chromosome condensation protein 1; fj77f08; guanine nucleotide-releasing protein; RanGEF; RCC1; rcc1.L; rcc1-a; rcc1-b; RCC1-I; Regulator of chromosome condensation; regulator of chromosome condensation 1; regulator of chromosome condensation 1 L homeolog; SNHG3-RCC1; SNHG3-RCC1 readthrough; wu:fj77f08; XELAEV_18012073mg; xrcc1; zgc:65796; zgc:77114 |
| Gene Symbols | RCC1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.