Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985484
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5484 Supplier Bioss Supplier No. BS3545R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Guanine-nucleotide releasing factor that promotes the exchange of Ran-bound GDP by GTP. Involved in the regulation of onset of chromosome condensation in the S phase. Binds both to the nucleosomes and double-stranded DNA. RCC1-Ran complex (together with other proteins) acts as a component of a signal transmission pathway that detects unreplicated DNA. Plays a key role in nucleo-cytoplasmic transport, mitosis and nuclear-envelope assembly.

Specifications

Antigen RCC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RCC1
Gene Accession No. P18754
Gene Alias 4931417M11Rik; AI326872; BOS_2368; Cell cycle regulatory protein; Chc1; chromosome condensation 1; Chromosome condensation protein 1; fj77f08; guanine nucleotide-releasing protein; Rcc1; RCC1-I; Regulator of chromosome condensation; regulator of chromosome condensation 1; SNHG3-RCC1; SNHG3-RCC1 readthrough; wu:fj77f08; zgc:65796; zgc:77114
Gene Symbols RCC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RCC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1104
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.